missing translation for 'onlineSavingsMsg'
Learn More
1-n-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals Product Code.: 11309645

1-n-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals

Product Code. 11309645
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11309645

Brand: Thermo Scientific Alfa Aesar L19087.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-n-Butyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used for various reactions such as hydrogenations, asymmetric hydrogenations proceed in higher enantioselectivity than in homogeneous phase. It is also used in Suzuki cross-coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Butyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used for various reactions such as hydrogenations, asymmetric hydrogenations proceed in higher enantioselectivity than in homogeneous phase. It is also used in Suzuki cross-coupling.

Solubility
Miscible with acetone, acetonitrile, ethyl acetate, isopropyl alcohol and methylene chloride. Immiscible with hexane, toluene and water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174501-65-6
Molecular Formula C8H15BF4N2
Molecular Weight (g/mol) 226.03
MDL Number MFCD03095449
InChI Key LSBXQLQATZTAPE-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborateShow MoreShow Less
PubChem CID 2734178
SMILES F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1

Specifications

Density 1.21
Flash Point None
Refractive Index 1.422
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Miscible with acetone,acetonitrile,ethyl acetate,isopropyl alcohol and methylene chloride. Immiscible with hexane,toluene and water.
IUPAC Name 1-butyl-3-methylimidazol-3-ium;tetrafluoroborate
Formula Weight 226.03
Percent Purity ≥98%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium tetrafluoroborate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241865 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberL19087.09,L19087.18
Lot Number10188880 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL19087.09,L19087.18
Lot Number10123568 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL19087.09,L19087.18
Lot Number10206687 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL19087.09,L19087.18
Lot Number10233948 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL19087.09,L19087.18
compliance-icons
Product Identifier
  • 1-n-Butyl-3-methylimidazoliumtetrafluoroborate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-n-Butyl-3-methylimidazoliumtetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-n-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.