missing translation for 'onlineSavingsMsg'
Learn More

1-n-Propylcyclohexanol, 98%, Thermo Scientific Chemicals

Product Code. 11430007
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5445-24-9
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00046364
InChI Key PYLPYOPJKOJRNP-UHFFFAOYSA-N
Synonym 1-propylcyclohexanol, 1-n-propylcyclohexanol, cyclohexanol, 1-propyl, 1-n-propyl cyclohexanol-1, propylcyclohexanol, 1-n-propyl-1-cyclohexanol, 1-n-propylcyclohexane-1-ol
PubChem CID 138504
IUPAC Name 1-propylcyclohexan-1-ol
SMILES CCCC1(O)CCCCC1

Specifications

Quantity 25 g
Formula Weight 142.24
Percent Purity 98%
Chemical Name or Material 1-n-Propylcyclohexanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'