missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Naphthalenemethanol, 98%, Thermo Scientific Chemicals
25.20 € - 68.50 €
Chemical Identifiers
CAS | 4780-79-4 |
---|---|
Molecular Formula | C11H10O |
Molecular Weight (g/mol) | 158.20 |
MDL Number | MFCD00004044 |
InChI Key | PBLNHHSDYFYZNC-UHFFFAOYSA-N |
Synonym | 1-naphthalenemethanol, 1-naphthylmethanol, 1-hydroxymethylnaphthalene, naphthalene-1-methanol, 1-naphthalene methanol, 1-menaphthyl alcohol, 1-naphthylmethyl alcohol, naphthalen-1-yl-methanol, naphthalen-1-yl methanol, 1-naphthylenemethanol |
PubChem CID | 20908 |
ChEBI | CHEBI:38137 |
IUPAC Name | naphthalen-1-ylmethanol |
SMILES | OCC1=C2C=CC=CC2=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10701271
|
Thermo Scientific Acros
180110020 |
2 g | Glass bottle |
25.20 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10144390
|
Thermo Scientific Acros
180110100 |
10 g | Glass bottle |
68.50 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4780-79-4 | |
158.20 | |
PBLNHHSDYFYZNC-UHFFFAOYSA-N | |
20908 | |
naphthalen-1-ylmethanol |
C11H10O | |
MFCD00004044 | |
1-naphthalenemethanol, 1-naphthylmethanol, 1-hydroxymethylnaphthalene, naphthalene-1-methanol, 1-naphthalene methanol, 1-menaphthyl alcohol, 1-naphthylmethyl alcohol, naphthalen-1-yl-methanol, naphthalen-1-yl methanol, 1-naphthylenemethanol | |
CHEBI:38137 | |
OCC1=C2C=CC=CC2=CC=C1 |
Specifications
4780-79-4 | |
100.0 | |
Yellow | |
Authentic | |
C11H10O | |
MFCD00004044 | |
1-naphthalenemethanol, 1-naphthylmethanol, 1-hydroxymethylnaphthalene, naphthalene-1-methanol, 1-naphthalene methanol, 1-menaphthyl alcohol, 1-naphthylmethyl alcohol, naphthalen-1-yl-methanol, naphthalen-1-yl methanol, 1-naphthylenemethanol | |
OCC1=C2C=CC=CC2=CC=C1 | |
158.20 | |
CHEBI:38137 | |
98% | |
1-Naphthalenemethanol, 98% |
97.5 | |
59°C to 62°C | |
301°C (715.0 mmHg) | |
98% | |
C10H7CH2OH | |
06, 667 | |
PBLNHHSDYFYZNC-UHFFFAOYSA-N | |
naphthalen-1-ylmethanol | |
20908 | |
158.2 | |
Fine Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 225-324-1
missing translation for 'rtecsNumber' : QJ8880000
missing translation for 'tsca' : TSCA
RUO – Research Use Only