Learn More
1-Naphthol, 99%, Thermo Scientific Chemicals
22.31 € - 246.23 €
Chemical Identifiers
CAS | 90-15-3 |
---|---|
Molecular Formula | C10H8O |
Molecular Weight (g/mol) | 144.173 |
MDL Number | MFCD00003930 |
InChI Key | KJCVRFUGPWSIIH-UHFFFAOYSA-N |
Synonym | 1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern |
PubChem CID | 7005 |
ChEBI | CHEBI:10319 |
IUPAC Name | naphthalen-1-ol |
SMILES | C1=CC=C2C(=C1)C=CC=C2O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11473824
|
Thermo Scientific Alfa Aesar
A11862.22 |
100 g |
22.31 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11483824
|
Thermo Scientific Alfa Aesar
A11862.36 |
500 g |
62.81 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493824
|
Thermo Scientific Alfa Aesar
A11862.0E |
2500 g |
246.23 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Naphthol is used as a precursor in the manufacturing of various azo dyes and pharmaceuticals such as nadolol. It is used as biomarkers. It is used in analytical chemistry as Molisch’s reagent (1-naphthol dissolved in ethanol) for checking the presence of carbohydrates. It plays an essential role with sodium hypobromite to detect the presence of arginine in proteins, which is called as Sakaguchi test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Naphthol is used as a precursor in the manufacturing of various azo dyes and pharmaceuticals such as nadolol. It is used as biomarkers. It is used in analytical chemistry as Molisch’s reagent (1-naphthol dissolved in ethanol) for checking the presence of carbohydrates. It plays an essential role with sodium hypobromite to detect the presence of arginine in proteins, which is called as Sakaguchi test.
Solubility
Soluble in benzene, chloroform, ether and ethanol.
Notes
Air and light sensitive. Store in a cool place. Incompatible with strong bases and strong oxidizing agents.
Chemical Identifiers
90-15-3 | |
144.173 | |
KJCVRFUGPWSIIH-UHFFFAOYSA-N | |
7005 | |
naphthalen-1-ol |
C10H8O | |
MFCD00003930 | |
1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern | |
CHEBI:10319 | |
C1=CC=C2C(=C1)C=CC=C2O |
Specifications
90-15-3 | |
1.28 | |
125°C (257°F) | |
C10H8O | |
UN2811 | |
Air and light sensitive | |
1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern | |
KJCVRFUGPWSIIH-UHFFFAOYSA-N | |
naphthalen-1-ol | |
7005 | |
144.17 | |
1-Naphthol |
95°C to 97°C | |
278°C to 280°C | |
Weak, Phenol-like | |
MFCD00003930 | |
1817321 | |
14,6383 | |
Soluble in benzene,chloroform,ether and ethanol. | |
C1=CC=C2C(=C1)C=CC=C2O | |
144.173 | |
CHEBI:10319 | |
99% |
Safety and Handling
GHS H Statement
H318-H302-H312-H335-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H318-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 201-969-4
missing translation for 'rtecsNumber' : QL2800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 1-Naphthol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : .alpha.-Naphthol
RUO – Research Use Only