missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoxyacetic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11334424
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2976-75-2
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.209
MDL Number MFCD00003927
InChI Key GHRYSOFWKRRLMI-UHFFFAOYSA-N
Synonym 1-naphthoxyacetic acid, 1-naphthyloxy acetic acid, 2-naphthalen-1-yloxy acetic acid, 1-naphthyloxyacetic acid, naphthyloxyacetic acid, 2-1-naphthyloxy acetic acid, unii-9qi2464poa, 1-naphthoxy acetic acid, naphthalen-1-yloxy acetic acid, acetic acid, 1-naphthalenyloxy
PubChem CID 76313
ChEBI CHEBI:44588
IUPAC Name 2-naphthalen-1-yloxyacetic acid
SMILES C1=CC=C2C(=C1)C=CC=C2OCC(=O)O

Specifications

Melting Point 193°C to 197°C
Quantity 25 g
Beilstein 1912165
Formula Weight 202.21
Percent Purity ≥98%
Chemical Name or Material 1-Naphthoxyacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'