missing translation for 'onlineSavingsMsg'
Learn More
1-Naphthyl isocyanate, 98%, Thermo Scientific Chemicals Product Code.: 11414160

1-Naphthyl isocyanate, 98%, Thermo Scientific Chemicals

Product Code. 11414160
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11414160

Brand: Thermo Scientific Alfa Aesar A17200.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Naphthyl isocyanate is used in the preparation of cationic aromatic urethane. It is useful for the study of cholangiolitic hepatotoxicity in laboratory animals. It is also employed as separate enantiomers of beta-blockers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl isocyanate is used in the preparation of cationic aromatic urethane. It is useful for the study of cholangiolitic hepatotoxicity in laboratory animals. It is also employed as separate enantiomers of beta-blockers.

Solubility
Miscible with alcohol, chloroform and diethyl ether.

Notes
Moisture and light sensitive. Incompatible with strong oxidizing agents and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-84-0
Molecular Formula C11H7NO
MDL Number MFCD00003881
InChI Key BDQNKCYCTYYMAA-UHFFFAOYSA-N
Synonym 1-naphthyl isocyanate, naphthalene, 1-isocyanato, alpha-naphthyl isocyanate, isocyanatonaphthalene, 1-naphthylisocyanate, 1-isocyanato-naphthalene, unii-5lh2p0691e, 1-isocyanatonapthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester
PubChem CID 66589
IUPAC Name 1-isocyanatonaphthalene

Specifications

Melting Point 4°C
Density 1.177
Boiling Point 269°C
Flash Point 135°C
Odor Irritating
Refractive Index 1.6335
Quantity 1 g
UN Number UN2206
Beilstein 742779
Sensitivity Moisture sensitive
Merck Index 14,6410
Solubility Information Miscible with alcohol,chloroform and diethyl ether.
SMILES O=C=NC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 169.18
Formula Weight 169.18
Percent Purity 98%
Chemical Name or Material 1-Naphthyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Naphthyl isocyanate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.