missing translation for 'onlineSavingsMsg'
Learn More
1-Naphthyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals Product Code.: 15431238

1-Naphthyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Product Code. 15431238
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15431238

Brand: Thermo Scientific Alfa Aesar H55091.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99747-74-7
Molecular Formula C11H7F3O3S
Molecular Weight (g/mol) 276.229
MDL Number MFCD00192340
InChI Key WQWUQDVFRYMMCY-UHFFFAOYSA-N
Synonym 1-naphthyl trifluoromethanesulfonate, 1-naphthyl triflate, 1-naphthyl-triflate, 1-naphthalenyl triflate, acmc-20aot0, 1-trifluoromethanesulfonyl naphthalene
PubChem CID 378884
IUPAC Name naphthalen-1-yl trifluoromethanesulfonate
SMILES C1=CC=C2C(=C1)C=CC=C2OS(=O)(=O)C(F)(F)F

Specifications

Color Red
Density 1.405 g/mL
Boiling Point 97°C to 98°C (0.3 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.52
Quantity 250 mg
UN Number UN3265
Formula Weight 276.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Naphthyl trifluoromethanesulfonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Naphthyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.