Learn More
1-Naphthylacetyl chloride, 95%, Thermo Scientific Chemicals
95.70 € - 464.00 €
Chemical Identifiers
CAS | 5121-00-6 |
---|---|
Molecular Formula | C12H9ClO |
Molecular Weight (g/mol) | 204.65 |
MDL Number | MFCD00175852 |
InChI Key | DSVAZLXLRDXHKO-UHFFFAOYSA-N |
Synonym | 2-1-naphthyl ethanoyl chloride, 2-naphthalen-1-yl acetyl chloride, 1-naphthylacetyl chloride, naphth-1-ylacetyl chloride, 1-naphthaleneacetyl chloride, naphth-1-yl acetyl chloride, napthylacetyl chloride, acmc-20aork, 1-naphthylacetylchloride, 1-naphthaleneacetylchloride |
PubChem CID | 2795386 |
IUPAC Name | 2-(naphthalen-1-yl)acetyl chloride |
SMILES | ClC(=O)CC1=CC=CC2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11395369
|
Thermo Scientific Alfa Aesar
H50386.03 |
1 g |
95.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11305379
|
Thermo Scientific Alfa Aesar
H50386.06 |
5 g |
464.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5121-00-6 | |
204.65 | |
DSVAZLXLRDXHKO-UHFFFAOYSA-N | |
2795386 | |
ClC(=O)CC1=CC=CC2=CC=CC=C12 |
C12H9ClO | |
MFCD00175852 | |
2-1-naphthyl ethanoyl chloride, 2-naphthalen-1-yl acetyl chloride, 1-naphthylacetyl chloride, naphth-1-ylacetyl chloride, 1-naphthaleneacetyl chloride, naphth-1-yl acetyl chloride, napthylacetyl chloride, acmc-20aork, 1-naphthylacetylchloride, 1-naphthaleneacetylchloride | |
2-(naphthalen-1-yl)acetyl chloride |
Specifications
5121-00-6 | |
130°C (0.6 mmHg) | |
C12H9ClO | |
MFCD00175852 | |
2-1-naphthyl ethanoyl chloride, 2-naphthalen-1-yl acetyl chloride, 1-naphthylacetyl chloride, naphth-1-ylacetyl chloride, 1-naphthaleneacetyl chloride, naphth-1-yl acetyl chloride, napthylacetyl chloride, acmc-20aork, 1-naphthylacetylchloride, 1-naphthaleneacetylchloride | |
ClC(=O)CC1=CC=CC2=CC=CC=C12 | |
204.65 | |
204.66 | |
1-Naphthylacetyl chloride |
1.23 | |
Pungent | |
1.621 | |
UN3265 | |
DSVAZLXLRDXHKO-UHFFFAOYSA-N | |
2-(naphthalen-1-yl)acetyl chloride | |
2795386 | |
95% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only