missing translation for 'onlineSavingsMsg'
Learn More

1-Nitroso-2-naphthol, 98%, Thermo Scientific Chemicals

44.42 € - 419.09 €

Chemical Identifiers

CAS 131-91-9
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00003884
InChI Key YXAOOTNFFAQIPZ-UHFFFAOYSA-N
Synonym 1-nitroso-2-naphthol, 2-naphthalenol, 1-nitroso, nitroso-beta-naphthol, 2-naphthol, 1-nitroso, 1-nitroso-2-naftol, zelen moridlova 4, zelen moridlova 4 czech, alpha-nitroso-beta-naphthol, 1-nitroso-2-naftol czech, unii-757i55u2qx
PubChem CID 8580
IUPAC Name 1-nitrosonaphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2N=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10123060
View Documents
Thermo Scientific Acros
151530250
25 g Glass bottle
44.42 €
25g
Please to purchase this item. Need a web account? Register with us today!
10183100
View Documents
Thermo Scientific Acros
151531000
100 g Glass bottle
131.66 €
100g
Please to purchase this item. Need a web account? Register with us today!
10489060
View Documents
Thermo Scientific Acros
151535000
500 g Glass bottle
419.09 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

131-91-9
173.17
YXAOOTNFFAQIPZ-UHFFFAOYSA-N
8580
C1=CC=C2C(=C1)C=CC(=C2N=O)O
C10H7NO2
MFCD00003884
1-nitroso-2-naphthol, 2-naphthalenol, 1-nitroso, nitroso-beta-naphthol, 2-naphthol, 1-nitroso, 1-nitroso-2-naftol, zelen moridlova 4, zelen moridlova 4 czech, alpha-nitroso-beta-naphthol, 1-nitroso-2-naftol czech, unii-757i55u2qx
1-nitrosonaphthalen-2-ol

Specifications

131-91-9
100.0
Brown
98%
ONC10H6OH
07, 712
1-nitroso-2-naphthol, 2-naphthalenol, 1-nitroso, nitroso-beta-naphthol, 2-naphthol, 1-nitroso, 1-nitroso-2-naftol, zelen moridlova 4, zelen moridlova 4 czech, alpha-nitroso-beta-naphthol, 1-nitroso-2-naftol czech, unii-757i55u2qx
YXAOOTNFFAQIPZ-UHFFFAOYSA-N
1-nitrosonaphthalen-2-ol
8580
98%
1-Nitroso-2-naphthol, 98%
97.5
104.0°C to 108.0°C
Authentic
C10H7NO2
MFCD00003884
15, 6728
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene,carbon,disulfide,caustic alkali solutions and glacial,acetic acid,slightly soluble in cold petroleum ether
C1=CC=C2C(=C1)C=CC(=C2N=O)O
173.17
173.17
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life.

GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a posit

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 205-043-0

missing translation for 'rtecsNumber' : QL4725000

missing translation for 'tsca' : TSCA

Product Identifier
  • 1-Nitroso-2-naphthol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only