missing translation for 'onlineSavingsMsg'
Learn More

1-Nonylamine, 97%, Thermo Scientific Chemicals

Product Code. 11473737
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Nonylamine is used as pharmaceutical intermediate. It is also used as a building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Nonylamine is used as pharmaceutical intermediate. It is also used as a building block.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-20-9
Molecular Formula C9H22ClN
Molecular Weight (g/mol) 179.73
MDL Number MFCD00008249
InChI Key IMGHCWMUWVHYCO-UHFFFAOYSA-N
Synonym nonylamine, n-nonylamine, 1-aminononane, 1-nonanamine, 1-nonylamine, unii-7l7h2ht4ok, 7l7h2ht4ok, nonyl-amine, n-nonyl amine, amine c9
PubChem CID 16215
IUPAC Name nonan-1-amine
SMILES [Cl-].CCCCCCCCC[NH3+]

Specifications

Melting Point -1°C
Density 0.784
Boiling Point 200°C to 202°C
Flash Point 62°C (143°F)
Odor Amine-like
Refractive Index 1.433
Quantity 100 g
UN Number UN2735
Beilstein 1733633
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 143.27
Percent Purity ≥98%
Chemical Name or Material 1-Nonylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.