Learn More
1-Octadecylamine, 97%, Thermo Scientific Chemicals
50.30 € - 137.00 €
Chemical Identifiers
CAS | 124-30-1 |
---|---|
Molecular Formula | C18H39N |
Molecular Weight (g/mol) | 269.52 |
MDL Number | MFCD00008159 |
InChI Key | REYJJPSVUYRZGE-UHFFFAOYSA-N |
Synonym | octadecylamine, 1-octadecanamine, stearylamine, 1-octadecylamine, octadecanamine, stearamine, 1-aminooctadecane, n-octadecylamine, n-stearylamine, armofilm |
PubChem CID | 15793 |
ChEBI | CHEBI:63866 |
IUPAC Name | octadecan-1-amine |
SMILES | CCCCCCCCCCCCCCCCCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348266
|
Thermo Scientific Alfa Aesar
L15458.14 |
25 g |
50.30 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358266
|
Thermo Scientific Alfa Aesar
L15458.22 |
100 g |
137.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Octadecylamine is used as asphalt emulsifier, lubricant and dispersants. It acts as a cationic surfactant and find applications is hair rinse bases, wood preservatives, textile softeners, dyeing auxiliaries and pigment grinding aids. It is used in boiler as an additive, which prevents the corrosion arises due to steam. It is a basic building block and used as an intermediate in the preparation of their amine derivatives, ethoxylates and amides. Metal oxide nano crystals having controlled size and shape are prepared by the thermal decomposition of metal nitrates with octadecylamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Octadecylamine is used as asphalt emulsifier, lubricant and dispersants. It acts as a cationic surfactant and find applications is hair rinse bases, wood preservatives, textile softeners, dyeing auxiliaries and pigment grinding aids. It is used in boiler as an additive, which prevents the corrosion arises due to steam. It is a basic building block and used as an intermediate in the preparation of their amine derivatives, ethoxylates and amides. Metal oxide nano crystals having controlled size and shape are prepared by the thermal decomposition of metal nitrates with octadecylamine.
Solubility
Soluble in chloroform, alcohol, ether, benzene and acetone.Insoluble in water.
Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
Chemical Identifiers
124-30-1 | |
269.52 | |
REYJJPSVUYRZGE-UHFFFAOYSA-N | |
15793 | |
octadecan-1-amine |
C18H39N | |
MFCD00008159 | |
octadecylamine, 1-octadecanamine, stearylamine, 1-octadecylamine, octadecanamine, stearamine, 1-aminooctadecane, n-octadecylamine, n-stearylamine, armofilm | |
CHEBI:63866 | |
CCCCCCCCCCCCCCCCCCN |
Specifications
124-30-1 | |
0.862 | |
148°C (298°F) | |
CH3(CH2)17NH2 | |
636111 | |
Soluble in chloroform,alcohol,ether,benzene and acetone.Insoluble in water. | |
CCCCCCCCCCCCCCCCCCN | |
269.52 | |
CHEBI:63866 | |
97% |
53°C to 55°C | |
180°C to 183°C (5 mmHg) | |
C18H39N | |
MFCD00008159 | |
octadecylamine, 1-octadecanamine, stearylamine, 1-octadecylamine, octadecanamine, stearamine, 1-aminooctadecane, n-octadecylamine, n-stearylamine, armofilm | |
REYJJPSVUYRZGE-UHFFFAOYSA-N | |
octadecan-1-amine | |
15793 | |
269.52 | |
1-Octadecylamine |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P262-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P310-P314-P330-P363-P501c
H302-H314-H318-H373
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 204-695-3
missing translation for 'rtecsNumber' : RG4150000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only