missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecylamine hydrochloride, Thermo Scientific Chemicals

Product Code. 11364088
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Octadecylamine hydrochloride is used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1838-08-0
Molecular Formula C18H40ClN
Molecular Weight (g/mol) 305.975
MDL Number MFCD00042018
InChI Key RNYJXPUAFDFIQJ-UHFFFAOYSA-N
Synonym octadecylamine hydrochloride, stearylamine hydrochloride, 1-octadecylamine hydrochloride, stearamine hydrochloride, unii-3v3v50q0sj, 1-octadecanamine, hydrochloride, octadecylamine, hydrochloride, acmc-1bnqe, octadecyl-ammonium chloride, c18h39n.hcl
PubChem CID 519552
IUPAC Name octadecan-1-amine;hydrochloride
SMILES CCCCCCCCCCCCCCCCCCN.Cl

Specifications

Melting Point 160°C
Quantity 5 g
Solubility Information Soluble in hot methanol.
Formula Weight 305.97
Physical Form Powder
Chemical Name or Material 1-Octadecylamine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'