missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octane Sulfonic Acid Sodium Salt, Ion Pair Chromatography, Fisher Chemical™
603.95 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Jar
Plastic powder jar
Description
C8H17NaO3S, CAS Number-5324-84-5, 1-octanesulfonate, sodium, sodium octylsulfonate, ipc-alks-8, 1-octanesulfonic acid, sodium salt, 1-octanesulfonic acid sodium salt, octyl sodium sulfonate, sodium octanesulfonate, sodium octane-1-sulfonate, sodium 1-octanesulfonate, unii-j46ez5jyb3, 100g, White
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 300°C |
Color | White |
pH | 5.5 to 7.5 |
Physical Form | Powder |
Quantity | 100 g |
IUPAC Name | sodium;octane-1-sulfonate |
Formula Weight | 216.28g/mol |
Grade | Chromatography |
Packaging | Plastic powder jar |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- 1-Octanesulfonic acid, sodium salt
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'