missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octanesulfonic acid, sodium salt monohydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | >300.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass Bottle |
Linear Formula | CH3(CH2)7SO3Na·H2O |
Quantity | 100 g |
Beilstein | 04,III,25 |
Solubility Information | Solubility in water: soluble. Other solubilities: 4g/l methanol |
Formula Weight | 234.3 |
Show More |
Safety and Handling

Product Identifier
- 1-Octanesulfonic acid, sodium salt monohydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-Octanesulfonic acid, sodium salt monohydrate, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction