missing translation for 'onlineSavingsMsg'
Learn More
1-Octanol, 99%, pure, Thermo Scientific Chemicals Product Code.: 10615202

1-Octanol, 99%, pure, Thermo Scientific Chemicals

Product Code. 10615202
2.5 L, Glass bottle
Click to view available options
Quantity:
250 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
This item is not returnable. View return policy

Product Code. 10615202

Brand: Thermo Scientific Acros 150630025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcoholShow MoreShow Less
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -16.0°C
Color Colorless
Density 0.8240g/mL
Boiling Point 195.0°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7OH
Refractive Index 1.4280 to 1.4310
Quantity 2.5 L
Beilstein 01, 418
Merck Index 15, 6840
Specific Gravity 0.824
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform,ether,and mineral oil,soluble in propylene glycol,insoluble in glycerol
Viscosity 9 mPa.s (20°C)
Formula Weight 130.23
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1-Octanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464583 Certificate TypeCertificate of Analysis Date15/07/2024 Catalog Number150630025,150632500,150630010,150630250
Lot NumberA0464368 Certificate TypeCertificate of Analysis Date08/07/2024 Catalog Number150630025,150632500,150630010,150630250
Lot NumberA0463719 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number150630025,150632500,150630010,150630250
Lot NumberA0460843 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number150630025,150632500,150630010,150630250
Lot NumberA0462306 Certificate TypeCertificate of Analysis Date26/04/2024 Catalog Number150630025,150632500,150630010,150630250
compliance-icons
Product Identifier
  • 1-Octanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Octanol, 99%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.