Learn More
1-Octanol, 99%, Thermo Scientific Chemicals
Description
1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.
Solubility
Miscible with ethanol, ether and propylene glycol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.

Specifications
Specifications
Melting Point | °C to 16°C |
Density | 0.825 |
Boiling Point | 193°C to 195°C |
Flash Point | 81°C (177°F) |
Assay Percent Range | 99% |
Linear Formula | CH3(CH2)7OH |
Refractive Index | 1.429 |
Quantity | 500 mL |
Beilstein | 1697461 |
Merck Index | 14,6751 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0462306 | Certificate of Analysis | 30/08/2024 | A15977.AP,A15977.0D,A15977.0F |
A0455836 | Certificate of Analysis | 08/03/2024 | A15977.AP,A15977.0D,A15977.0F |
A0457508 | Certificate of Analysis | 08/03/2024 | A15977.AP,A15977.0D,A15977.0F |
A0453932 | Certificate of Analysis | 10/08/2023 | A15977.AP,A15977.0D,A15977.0F |
A0454684 | Certificate of Analysis | 10/08/2023 | A15977.AP,A15977.0D,A15977.0F |
Safety and Handling

- 1-Octanol
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : 1-octanol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.