missing translation for 'onlineSavingsMsg'
Learn More

1-Octene, 99+%

Product Code. 10772951 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
Unit Size:
1 L
10 L
100mL
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10772951

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-66-0 | C8H16 | 112.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-66-0
Molecular Formula C8H16
Molecular Weight (g/mol) 112.21
MDL Number MFCD00009548
InChI Key KWKAKUADMBZCLK-UHFFFAOYSA-N
Synonym 1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene
PubChem CID 8125
ChEBI CHEBI:46708
IUPAC Name oct-1-ene
SMILES CCCCCCC=C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -102.0°C
Density 0.7150g/mL
Boiling Point 121.0°C to 123.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5CH=CH2
Refractive Index 1.4080 to 1.4100
Quantity 100 mL
Beilstein 01, 221
Merck Index 15, 1766
Specific Gravity 0.715
Solubility Information Solubility in water: 4.1 mg/l (25°C). Other solubilities: soluble in alcohol, acetone, ether, petroleum and, coal-tar solvents, soluble in hydrocarbon solvents
Viscosity 0.38 mm2/s (40°C)
Formula Weight 112.21
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1-Octene, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Octene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contains : 1-Octene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.