Learn More
1-Octylamine, 99%, Thermo Scientific Chemicals
24.32 € - 77.18 €
Chemical Identifiers
CAS | 111-86-4 |
---|---|
Molecular Formula | C8H19N |
Molecular Weight (g/mol) | 129.247 |
MDL Number | MFCD00008247 |
InChI Key | IOQPZZOEVPZRBK-UHFFFAOYSA-N |
Synonym | octylamine, n-octylamine, 1-aminooctane, 1-octanamine, 1-octylamine, caprylamine, caprylylamine, armeen 8, n-octylamine, mono, armeen 8d |
PubChem CID | 8143 |
ChEBI | CHEBI:7728 |
IUPAC Name | octan-1-amine |
SMILES | CCCCCCCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11421237
|
Thermo Scientific Alfa Aesar
B24193.22 |
100 g |
24.32 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11431237
|
Thermo Scientific Alfa Aesar
B24193.36 |
500 g |
77.18 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Octylamine is used as a precursor to prepare chemicals. It finds application in functional fluids and as a laboratory reagent. Further, it is used as a reactant in the preparation of 2H-indazoles and 1H-indazolones, which are used as myeloperoxidase(MPO) inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Octylamine is used as a precursor to prepare chemicals. It finds application in functional fluids and as a laboratory reagent. Further, it is used as a reactant in the preparation of 2H-indazoles and 1H-indazolones, which are used as myeloperoxidase(MPO) inhibitors.
Solubility
Miscible with ethanol, benzene and ethyl acetate. Slightly miscible with water.
Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides and acid chlorides.
Chemical Identifiers
111-86-4 | |
129.247 | |
IOQPZZOEVPZRBK-UHFFFAOYSA-N | |
8143 | |
octan-1-amine |
C8H19N | |
MFCD00008247 | |
octylamine, n-octylamine, 1-aminooctane, 1-octanamine, 1-octylamine, caprylamine, caprylylamine, armeen 8, n-octylamine, mono, armeen 8d | |
CHEBI:7728 | |
CCCCCCCCN |
Specifications
111-86-4 | |
0.78 | |
60°C (144°F) | |
1.429 | |
UN2735 | |
Air Sensitive | |
Miscible with ethanol,benzene and ethyl acetate. Slightly miscible with water. | |
CCCCCCCCN | |
129.247 | |
CHEBI:7728 | |
99% |
-1°C | |
178°C to 179°C | |
C8H19N | |
MFCD00008247 | |
1679227 | |
octylamine, n-octylamine, 1-aminooctane, 1-octanamine, 1-octylamine, caprylamine, caprylylamine, armeen 8, n-octylamine, mono, armeen 8d | |
IOQPZZOEVPZRBK-UHFFFAOYSA-N | |
octan-1-amine | |
8143 | |
129.25 | |
1-Octylamine |
Safety and Handling
GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H301+H311-H314-H332-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: I
missing translation for 'einecsNumber' : 203-916-0
missing translation for 'rtecsNumber' : RG8050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 1-Octylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 1-Octanamine
RUO – Research Use Only