missing translation for 'onlineSavingsMsg'
Learn More

1-Octylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11344346
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Octylboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octylboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28741-08-4
Molecular Formula C8H19BO2
Molecular Weight (g/mol) 158.048
MDL Number MFCD01074560
InChI Key GKFRVXOKPXCXAK-UHFFFAOYSA-N
Synonym n-octylboronic acid, 1-octylboronic acid, boronic acid, octyl, 1-n-octaneboronic acid, boronic acid, b-octyl, caprylboronic acid, acmc-1cbf1, n-octaneboronic acid, b-octyl-boronic acid
PubChem CID 5195177
IUPAC Name octylboronic acid
SMILES B(CCCCCCCC)(O)O

Specifications

Melting Point 81°C to 85°C
Quantity 5 g
Beilstein 1739759
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 158.05
Percent Purity 97%
Chemical Name or Material 1-Octylboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'