missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Oleoyl lysophosphatidic acid sodium salt, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Description
1-Oleoyl lysophosphatidic acid sodium salt is an endogenous agonist of the lysophospholipid receptors LPA1 and LPA2. 1-Oleoyl lysophosphatidic acid sodium salt inhibits differentiation of neural stem cells (NSCs) into neurons.
Note: This product is typically prepared in PBS, pH 7.2, at a concentration of up to approximately 4 mg/mL.
Specifications
Specifications
CAS | 325465-93-8 |
Molecular Formula | C21H40NaO7P |
Quantity | 1 mg |
Synonym | 1-oleoyl lysophosphatidic acid sodium salt, c21h40o7p.na, bml3-f05, d-glycerol 1-phosphoric acid sodium 3-oleate salt, 1-oleoyl-2-hydroxy-sn-glycero-3-phosphate sodium salt, 1-o-9z-octadecenoyl-sn-glyceryl-3-phosphoric acid sodium salt, sodium r,z-2-hydroxy-3-oleoyloxy propyl hydrogenphosphate, sodium 2r-3-hydrogen phosphonatooxy-2-hydroxypropyl 9z-octadec-9-enoate |
InChI Key | XGRLSUFHELJJAB-JGSYTFBMSA-M |
SMILES | CCCCCCCCC=CCCCCCCCC(=O)OCC(COP(=O)(O)[O-])O.[Na+] |
IUPAC Name | sodium;[(2R)-2-hydroxy-3-[(Z)-octadec-9-enoyl]oxypropyl] hydrogen phosphate |
Molecular Weight (g/mol) | 458.508 |
PubChem CID | 44159357 |
Formula Weight | 458.5 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'