missing translation for 'onlineSavingsMsg'
Learn More

1-Pentene, 97%

Product Code. 11358661 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
25mL
500mL
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 11358661

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 109-67-1 | C5H10 | 70.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-67-1
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00003567
InChI Key YWAKXRMUMFPDSH-UHFFFAOYSA-N
Synonym 1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
PubChem CID 8004
IUPAC Name pent-1-ene
SMILES CCCC=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -165.0°C
Color Colorless
Density 0.6400g/mL
Boiling Point 30.0°C
Flash Point −28°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3CH2CH2CH=CH2
Refractive Index 1.3704 to 1.3724
Quantity 2.5 L
Beilstein 01,21
Merck Index 15,7234
Specific Gravity 0.64
Solubility Information Solubility in water: 0.15g/L (20°C). Other solubilities: miscible with alcohol, ether and beznene, soluble in most organic solvents
Viscosity 0.202 mPa.s (20°C)
Formula Weight 70.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Pentene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Pentene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 1
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : 1-Pentene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.