missing translation for 'onlineSavingsMsg'
Learn More

1-Pentylamine, 98%, Thermo Scientific Chemicals

32.16 € - 404.01 €

Chemical Identifiers

CAS 110-58-7
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008236
InChI Key DPBLXKKOBLCELK-UHFFFAOYSA-N
Synonym amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine
PubChem CID 8060
ChEBI CHEBI:74848
IUPAC Name pentan-1-amine
SMILES CCCCCN
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11449163
View Documents
Thermo Scientific Alfa Aesar
A11674.14
25 g
32.16 €
25g
Please to purchase this item. Need a web account? Register with us today!
11459163
View Documents
Thermo Scientific Alfa Aesar
A11674.22
100 g
91.96 €
100g
Please to purchase this item. Need a web account? Register with us today!
11469163
View Documents
Thermo Scientific Alfa Aesar
A11674.36
500 g
404.01 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.

Solubility
Soluble in alcohol, ether.

Notes
Air Sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

110-58-7
87.166
DPBLXKKOBLCELK-UHFFFAOYSA-N
8060
pentan-1-amine
C5H13N
MFCD00008236
amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine
CHEBI:74848
CCCCCN

Specifications

110-58-7
0.751
−1°C (30°F)
C5H13N
MFCD00008236
505953
14,598
Soluble in alcohol,ether.
CCCCCN
87.166
CHEBI:74848
98%
-55°C
103°C to 105°C
Amine-like
1.411
UN1106
Air Sensitive
amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine
DPBLXKKOBLCELK-UHFFFAOYSA-N
pentan-1-amine
8060
87.17
1-Pentylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H331-H314-H318-H302-H312
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H312-H314-H331-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYLAMINES

missing translation for 'einecsNumber' : 203-780-2

missing translation for 'rtecsNumber' : SC0300000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 1-Pentylamine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : 1-Pentanamine

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.