missing translation for 'onlineSavingsMsg'
Learn More

1-Pentyne, 99%

Product Code. 10105403 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10105403

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-19-0 | C5H8 | 68.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-19-0
Molecular Formula C5H8
Molecular Weight (g/mol) 68.12
MDL Number MFCD00009469
InChI Key IBXNCJKFFQIKKY-UHFFFAOYSA-N
Synonym 1-pentyne, propylacetylene, pentyne, acetylene, propyl, pentin, pentyn, 1-n-pentyne, pent-1-yn-1-yl, acmc-1cuk3
PubChem CID 12309
IUPAC Name pent-1-yne
SMILES CCCC#C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -106°C to 105°C
Color Colorless to Yellow
Density 0.6900g/mL
Boiling Point 40.0°C
Flash Point <−34°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2C≡CH
Refractive Index 1.3840 to 1.3880
Quantity 5 g
Specific Gravity 0.69
Formula Weight 68.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Pentyne, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Pentyne
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 1-Pentyne

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.