missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1,2-ethanediol, 97%, Thermo Scientific Chemicals

Product Code. 10550594
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-56-1
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00003546
InChI Key PWMWNFMRSKOCEY-UHFFFAOYSA-N
Synonym 1-phenyl-1,2-ethanediol, styrene glycol, phenylethylene glycol, phenylethanediol, phenyl glycol, 1,2-ethanediol, 1-phenyl, phenyl-1,2-ethanediol, styrolyl alcohol, fenylglycol, 1-phenylethylene glycol
PubChem CID 7149
IUPAC Name 1-phenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(CO)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 61°C to 67°C
Color White to Beige
Boiling Point 272°C to 274°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula HOCH2CH(C6H5)OH
Quantity 100 g
Beilstein 06, 907
Merck Index 15, 8991
Formula Weight 138.17
Percent Purity 97%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 1-Phenyl-1, 2-ethanediol
Show More Show Less
Product Identifier
  • 1-Phenyl-1,2-ethanediol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.