Learn More
1-Phenyl-1,4-pentanedione, 96%, Thermo Scientific Chemicals
38.79 € - 120.60 €
Chemical Identifiers
CAS | 583-05-1 |
---|---|
Molecular Formula | C11H12O2 |
Molecular Weight (g/mol) | 176.215 |
MDL Number | MFCD00092889 |
InChI Key | RBLXWIPBPPVLPU-UHFFFAOYSA-N |
Synonym | 1-phenyl-1,4-pentanedione, 1-phenyl-pentane-1,4-dione, 1,4-pentanedione, 1-phenyl, acmc-20aosh, 1-phenyl-1,4-pentandione, 5-phenyl-2,5-pentanedione, benzil-related compound, 41, 1-phenyl pentane-1,4-dione |
PubChem CID | 136371 |
IUPAC Name | 1-phenylpentane-1,4-dione |
SMILES | CC(=O)CCC(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11367936
|
Thermo Scientific Alfa Aesar
L13151.03 |
1 g |
38.79 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377936
|
Thermo Scientific Alfa Aesar
L13151.06 |
5 g |
120.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Phenyl-1,4-pentanedione is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Phenyl-1,4-pentanedione is used as an organic chemical synthesis intermediate.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
583-05-1 | |
176.215 | |
RBLXWIPBPPVLPU-UHFFFAOYSA-N | |
136371 | |
CC(=O)CCC(=O)C1=CC=CC=C1 |
C11H12O2 | |
MFCD00092889 | |
1-phenyl-1,4-pentanedione, 1-phenyl-pentane-1,4-dione, 1,4-pentanedione, 1-phenyl, acmc-20aosh, 1-phenyl-1,4-pentandione, 5-phenyl-2,5-pentanedione, benzil-related compound, 41, 1-phenyl pentane-1,4-dione | |
1-phenylpentane-1,4-dione |
Specifications
583-05-1 | |
C11H12O2 | |
MFCD00092889 | |
1-phenyl-1,4-pentanedione, 1-phenyl-pentane-1,4-dione, 1,4-pentanedione, 1-phenyl, acmc-20aosh, 1-phenyl-1,4-pentandione, 5-phenyl-2,5-pentanedione, benzil-related compound, 41, 1-phenyl pentane-1,4-dione | |
RBLXWIPBPPVLPU-UHFFFAOYSA-N | |
1-phenylpentane-1,4-dione | |
136371 | |
96% |
158°C to 162°C (12 mmHg) | |
1.533 | |
908230 | |
Not miscible in water. | |
CC(=O)CCC(=O)C1=CC=CC=C1 | |
176.215 | |
176.22 | |
1-Phenyl-1,4-pentanedione |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.