missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Phenylethanol, 97%, Thermo Scientific Chemicals

69.30 € - 731.00 €

Chemical Identifiers

CAS 98-85-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004508
InChI Key WAPNOHKVXSQRPX-UHFFFAOYSA-N
Synonym methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene
PubChem CID 7409
ChEBI CHEBI:669
IUPAC Name 1-phenylethanol
SMILES CC(C1=CC=CC=C1)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11444934
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13837.36
500 g
69.30 €
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11451478
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13837.0E
2500 g
266.00 €
2500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11461478
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13837.0C
10,000 g
731.00 €
10000g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Arthrobacter sp. is able to grow with (+ I-,(-)-1-phenylethanol or the racemic mixture as sole source of carbon. Growth is most rapid with the (-)-isomer, doubling time 12 h. Lipases show good activity and, in some cases, improved enantioselectivity when employed in pure ionic liquids for dynamic kinetic resolution of 1-phenylethanol by transesterification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The Arthrobacter sp. is able to grow with (+ I-,(-)-1-phenylethanol or the racemic mixture as sole source of carbon. Growth is most rapid with the (-)-isomer, doubling time 12 h. Lipases show good activity and, in some cases, improved enantioselectivity when employed in pure ionic liquids for dynamic kinetic resolution of 1-phenylethanol by transesterification.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
Specifications

Chemical Identifiers

98-85-1
122.167
WAPNOHKVXSQRPX-UHFFFAOYSA-N
7409
1-phenylethanol
C8H10O
MFCD00004508
methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene
CHEBI:669
CC(C1=CC=CC=C1)O

Specifications

98-85-1
1.012
C8H10O
UN2937
Insoluble in water.
CC(C1=CC=CC=C1)O
122.167
CHEBI:669
97%
19°C to 20°C
203°C to 205°C
MFCD00004508
methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene
WAPNOHKVXSQRPX-UHFFFAOYSA-N
1-phenylethanol
7409
122.17
(+/-)-1-Phenylethanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H227-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.

P210-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P370+P378q-P501c

H227-H302-H319

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALPHA-METHYLBENZYL ALCOHOL, LIQUID

missing translation for 'einecsNumber' : 202-707-1

missing translation for 'rtecsNumber' : DO9275000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only