missing translation for 'onlineSavingsMsg'
Learn More
1-Propanol, 99.5%, Thermo Scientific Chemicals Product Code.: 10003493

1-Propanol, 99.5%, Thermo Scientific Chemicals

Product Code. 10003493
100 mL
Click to view available options
Quantity:
100 mL
1 L
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10003493

Brand: Thermo Scientific Acros 396941000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcoholShow MoreShow Less
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127.0°C
CAS Min % 99.4
CAS Max % 100.0
Density 0.8000g/mL
Boiling Point 97.0°C
Quantity 100 mL
Flash Point 15°C
Infrared Spectrum Authentic
Physical Form Liquid
Refractive Index 1.3840 to 1.3860
Assay Percent Range 99.5%
Residue after Evaporation 0.001 % max.
Specific Gravity 0.8
Linear Formula CH3CH2CH2OH
Viscosity 2.2 mPa.s (20°C)
Water 0.005% Max. (Coulometric)
Merck Index 15, 7955
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents
Formula Weight 60.1
Percent Purity 99.5%
Grade Extra Dry
Chemical Name or Material 1-Propanol, Extra Dry
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2415020 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number396941000,396940010
Lot Number2495647 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number396941000,396940010
Lot Number1903792 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number396941000,396940010
Lot Number2047535 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number396941000,396940010
Lot Number2031628 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number396941000,396940010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Propanol, 99.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.