missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, puriss. p.a., ACS Reagent, ≥99.5% (GC), Honeywell™ Riedel-de Haën™
BIOPHARMA

Product Code. 15623990
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Poly Bottle

Puriss. p.a., Reag. Ph. Eur.,≥99.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127°C
Color Colorless
Density 0.804g/mL at 25°C
Boiling Point 97°C
Quantity 6 x 1 L
Explosion Limit 0.137
Flash Point 22°C (71.6°F)
Ignition Point 700°F
Odor Alcohol-like
Packaging Poly Bottle
Refractive Index n20/D 1.384
Linear Formula CH3CH2CH2OH
Vapor Pressure 14.9mmHg (20°C), 10mmHg (147°C)
UN Number UN1274
Beilstein 1098242
Solubility Information Soluble in water and most organic solvents
Formula Weight 60.1g/mol
Percent Purity ≥99.5% (GC)
Chemical Name or Material 1-Propanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : propan-1-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.