missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propanol, puriss. p.a., Honeywell
Click to view available options
Quantity:
1 L
2.5 L
160 kg
Specifications
Density | 0.804g/mL at 25°C |
Quantity | 2.5 L |
Explosion Limit | 0.137 |
Flash Point | 22°C (71.6°F) |
Ignition Point | 700°F |
Linear Formula | CH3CH2CH2OH |
Vapor Pressure | 14.9 mmHg (20°C), 10 mmHg (147°C) |
UN Number | UN1274 |
Beilstein | 1098242 |
Formula Weight | 60.1g/mol |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'