missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propanol, purum, ≥99% (GC), Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Melting Point | -127° |
Density | 0.804g/mL at 25°C |
Boiling Point | 97° |
Quantity | 6 x 1 L |
Explosion Limit | 0.137 |
Flash Point | 22°C (71.6°F) |
Ignition Point | 700°F |
Packaging | Poly Bottle |
Physical Form | Liquid |
Linear Formula | CH3CH2CH2OH |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-Propanol, purum, ≥99% (GC), Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction