missing translation for 'onlineSavingsMsg'
Learn More

1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

176.00 € - 420.00 €

Chemical Identifiers

CAS 16466-97-0, 109-99-9
Molecular Formula C3H3BrMg
Molecular Weight (g/mol) 143.27
MDL Number MFCD00216588
InChI Key YVJBCTFVENNIOV-UHFFFAOYSA-M
Synonym 1-propynylmagnesium bromide, 1-propynyl magnesium bromide, bromo prop-1-yn-1-yl magnesium, 1-propynylmagnesium bromide solution, propynylmagnesiumbromide, propynyl magnesium bromide, prop-1-ynylmagnesium bromide, prop-1-inyl magnesium bromide, prop-1-ynyl-magnesium bromide, propyn-1-yl magnesium bromide, grignard reagent
PubChem CID 3608069
IUPAC Name magnesium;prop-1-yne;bromide
SMILES CC#C[Mg]Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10396724
View Documents
Thermo Scientific Acros
426071000
100 mL AcroSeal™ Glass bottle
176.00 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10796275
View Documents
Thermo Scientific Acros
426078000
800 mL AcroSeal™ Glass bottle
420.00 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16466-97-0, 109-99-9
143.27
YVJBCTFVENNIOV-UHFFFAOYSA-M
3608069
CC#C[Mg]Br
C3H3BrMg
MFCD00216588
1-propynylmagnesium bromide, 1-propynyl magnesium bromide, bromo prop-1-yn-1-yl magnesium, 1-propynylmagnesium bromide solution, propynylmagnesiumbromide, propynyl magnesium bromide, prop-1-ynylmagnesium bromide, prop-1-inyl magnesium bromide, prop-1-ynyl-magnesium bromide, propyn-1-yl magnesium bromide, grignard reagent
magnesium;prop-1-yne;bromide

Specifications

16466-97-0, 109-99-9
65.0°C to 67.0°C
C3H3BrMg
0.94
YVJBCTFVENNIOV-UHFFFAOYSA-M
magnesium;prop-1-yne;bromide
3608069
1-Propynylmagnesium bromide
0.9400g/mL
−1°C
MFCD00216588
1-propynylmagnesium bromide, 1-propynyl magnesium bromide, bromo prop-1-yn-1-yl magnesium, 1-propynylmagnesium bromide solution, propynylmagnesiumbromide, propynyl magnesium bromide, prop-1-ynylmagnesium bromide, prop-1-inyl magnesium bromide, prop-1-ynyl-magnesium bromide, propyn-1-yl magnesium bromide, grignard reagent
CC#C[Mg]Br
143.27
143.26
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
May form explosive peroxides.
Reacts violentl

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

Product Identifier
  • 1-Propynylmagnesium bromide, 0.5M solution in THF

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H261-In contact with water releases flammable gases.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.