missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrenecarboxaldehyde, 98+%, Thermo Scientific Chemicals

Product Code. 11363016
Click to view available options
Quantity:
5 g
25 g

1-Pyrenecarboxaldehyde is a important intermediate used in agrochemical, pharmaceutical and dyestuff field. It is used as a highly polarity-sensitive fluorophore for SNP typing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrenecarboxaldehyde is a important intermediate used in agrochemical, pharmaceutical and dyestuff field. It is used as a highly polarity-sensitive fluorophore for SNP typing.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3029-19-4
Molecular Formula C17H10O
Molecular Weight (g/mol) 230.266
MDL Number MFCD00004139
InChI Key RCYFOPUXRMOLQM-UHFFFAOYSA-N
Synonym 1-pyrenecarboxaldehyde, 1-formylpyrene, 1-pyrenealdehyde, pyrenecarboxaldehyde, 3-formylpyrene, 3-pyrenealdehyde, 3-pyrenylaldehyde, pyrene-1-aldehyde, 3-pyrenecarboxaldehyde, 1-pyrenecarbaldehyde
PubChem CID 232848
IUPAC Name pyrene-1-carbaldehyde
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C=O

Specifications

Melting Point 125°C to 128°C
Quantity 25 g
Beilstein 1874889
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 230.27
Percent Purity 99%
Chemical Name or Material 1-Pyrenecarboxaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.