missing translation for 'onlineSavingsMsg'
Learn More

1-(pyrid-3-ylmethyl)piperidine-4-carboxylic acid dihydrochloride, 97%, Thermo Scientific™

Product Code. 10575944
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 946409-40-1
Molecular Formula C12H18Cl2N2O2
Molecular Weight (g/mol) 293.188
MDL Number MFCD06801260
InChI Key FLINLLCJYJQIIY-UHFFFAOYSA-N
Synonym 1-pyridin-3-ylmethyl piperidine-4-carboxylic acid dihydrochloride, 1-pyrid-3-ylmethyl piperidine-4-carboxylic acid dihydrochloride, 1-pyridin-3-yl methyl piperidine-4-carboxylic acid dihydrochloride, 1-pyridin-3-yl methyl piperidine-4-carboxylic acid-hydrogen chloride 1/2
PubChem CID 42935493
IUPAC Name 1-(pyridin-3-ylmethyl)piperidine-4-carboxylic acid;dihydrochloride
SMILES C1CN(CCC1C(=O)O)CC2=CN=CC=C2.Cl.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 293.19
Percent Purity 95%
Chemical Name or Material 1-(Pyrid-3-ylmethyl)piperidine-4-carboxylic acid dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.