missing translation for 'onlineSavingsMsg'
Learn More
1-(Pyridin-3-ylmethyl)-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific™ Product Code.: 15572303

1-(Pyridin-3-ylmethyl)-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 15572303
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15572303

Brand: Thermo Scientific Acros 449970050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 864754-21-2
Molecular Formula C15H20BN3O2
Molecular Weight (g/mol) 285.15
MDL Number MFCD08706029
InChI Key LVLQAPQDEZOFQU-UHFFFAOYSA-N
Synonym 3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl methyl pyridine, 1-pyridin-3-ylmethyl-1h-pyrazole-4-boronic acid, pinacol ester, 3-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrazol-1-ylmethyl-pyridine, 1-3-pyridylmethyl-1h-pyrazole-4-boronic acid pinacol ester, 3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl methyl pyridine, 3-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl methyl pyridine, pubchem18546, 1-pyridin-3-ylmethyl-1h-pyrazole-4-boronic acidShow MoreShow Less
PubChem CID 17999185
SMILES CC1(C)OB(OC1(C)C)C1=CN(CC2=CN=CC=C2)N=C1

Specifications

Melting Point 102.0°C to 106.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
IUPAC Name 3-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]methyl]pyridine
Formula Weight 285.16
Percent Purity 97%
Chemical Name or Material 1-(Pyridin-3-ylmethyl)-1H-pyrazole-4-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 1-(Pyridin-3-ylmethyl)-1H-pyrazole-4-boronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(Pyridin-3-ylmethyl)-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.