Learn More
1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%, Thermo Scientific Chemicals
1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%, C5H12N2S2, CAS Number-5108-96-3
44.02 € - 516.57 €
Chemische Identifikatoren
CAS | 5108-96-3 |
---|---|
Molecular Formula | C5H12N2S2 |
Molecular Weight (g/mol) | 164.28 |
MDL Number | MFCD00012720 |
InChI Key | VSWDORGPIHIGNW-UHFFFAOYSA-M |
Synonym | ammonium pyrrolidyldithiocarbamate, apdc, ammonium 1-pyrrolidinecarbodithioate, ammonium tetramethylenedithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium 1-pyrrolidinedithiocarboxylate, 1-pyrrolidinecarbodithioic acid, ammonium salt, 1-pyrrolidinecarbodithioic acid ammonium salt, pyrrolidinedithiocarbamate ammonium |
PubChem CID | 4311638 |
IUPAC Name | pyrrolidine-1-carbodithioate |
SMILES | C1CCN(C1)C(=S)[S-] |
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
10572371
|
Thermo Scientific Acros
162050250 |
25 g | Glass bottle |
44.02 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10081870
|
Thermo Scientific Acros
162051000 |
100 g | Glass bottle |
143.72 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10297353
|
Thermo Scientific Acros
162055000 |
500 g | Glass bottle |
516.57 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
5108-96-3 | |
164.28 | |
VSWDORGPIHIGNW-UHFFFAOYSA-M | |
4311638 | |
C1CCN(C1)C(=S)[S-] |
C5H12N2S2 | |
MFCD00012720 | |
ammonium pyrrolidyldithiocarbamate, apdc, ammonium 1-pyrrolidinecarbodithioate, ammonium tetramethylenedithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium 1-pyrrolidinedithiocarboxylate, 1-pyrrolidinecarbodithioic acid, ammonium salt, 1-pyrrolidinecarbodithioic acid ammonium salt, pyrrolidinedithiocarbamate ammonium | |
pyrrolidine-1-carbodithioate |
Spezifikation
5108-96-3 | |
100.0 | |
White | |
98% | |
MFCD00012720 | |
Solubility in water: 50g/L (20°C). Other solubilities: soluble in ethanol and chloroform | |
C1CCN(C1)C(=S)[S-] | |
164.28 | |
164.28 | |
Powder |
97.5 | |
150.0°C to 155.0°C | |
Authentic | |
C5H12N2S2 | |
ammonium pyrrolidyldithiocarbamate, apdc, ammonium 1-pyrrolidinecarbodithioate, ammonium tetramethylenedithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium 1-pyrrolidinedithiocarboxylate, 1-pyrrolidinecarbodithioic acid, ammonium salt, 1-pyrrolidinecarbodithioic acid ammonium salt, pyrrolidinedithiocarbamate ammonium | |
VSWDORGPIHIGNW-UHFFFAOYSA-M | |
pyrrolidine-1-carbodithioate | |
4311638 | |
98% | |
1-Pyrrolidinecarbodithioic acid, ammonium salt |
Sicherheit und Handhabung
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 225-834-4
- 1-Pyrrolidinecarbodithioic acid, ammonium salt
Signalwort
- Warning
Gefahrenkategorie
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Gefahrenhinweis
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Sicherheitshinweis
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.