Learn More
Microspec™ 1 Roll Autoclave Tape
Autoclavable tape roll
Brand:  Microspec™ X2
 View more versions of this product
Product Code. 16695482
  • 8.10 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Autoclavable
  • Different sizes available
Specifications
Specifications
Autoclavable
121°C
Non-sterile
25 mm x 52 m
1