Learn More
Thermo Scientific™ 1-Step™ Ultra TMB-Blotting Solution
Thermo Scientific™ 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting and immunohistochemistry.
Brand: Thermo Scientific™ 37574
241.30 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting and immunohistochemistry.
Additional Information and Online Ordering
Thermo Scientific 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting.
This precipitating, colorimetric Western blot substrate for HRP provides high sensitivity, increased signal-to-noise ratio and low background compared to many other chromogenic substrates. The blotting solution contains soluble TMB (3,3',5,5'-tetramethylbenzidine), which reacts very quickly with horseradish peroxidase enzyme to produce an insoluble dark blue precipitate that does not fade or flake. The substrate is compatible with both nitrocellulose and PVDF membranes. The blotting solution is supplied ready to use with no mixing required.
Features of 1-Step Ultra TMB-Blotting Solution:
• Fast—protein bands visible in less than one minute
• Fade-resistant—protein bands stable after membrane drying
• Sensitive—detection limit similar to Pierce ECL Western Blotting Substrate
• Chromogenic—no special equipment needed for visualization; produces dark blue bands
• Ready to use—no organic solvents are required to dissolve; no dilution necessary for use
Applications:
• Chromogenic Western blot detection
• Nitrocellulose membrane blots
• PVDF membrane blots

Specifications
Substrate | |
Liquid | |
1-Step™ | |
TMB |
Colorimetric | |
Nitrocellulose, PVDF | |
250 mL | |
Upon receipt store product at 4°C. Product is shipped at ambient temperature. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.