missing translation for 'onlineSavingsMsg'
Learn More

(1-Thieno[3,2-d]pyrimidin-4-ylpiperid-4-yl)methanol, 97%, Thermo Scientific™

Product Code. 11366855
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 910037-26-2
Molecular Formula C12H15N3OS
Molecular Weight (g/mol) 249.332
MDL Number MFCD09065024
InChI Key UXMVQNYPOJJGNA-UHFFFAOYSA-N
Synonym 1-thieno 3,2-d pyrimidin-4-ylpiperid-4-yl methanol, 1-thieno 3,2-d pyrimidin-4-yl piperidin-4-yl methanol, 4-piperidinemethanol,1-thieno 3,2-d pyrimidin-4-yl, 1-thiopheno 2,3-e pyrimidin-4-yl-4-piperidyl methan-1-ol
PubChem CID 24229707
IUPAC Name (1-thieno[3,2-d]pyrimidin-4-ylpiperidin-4-yl)methanol
SMILES C1CN(CCC1CO)C2=NC=NC3=C2SC=C3

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 249.34
Percent Purity 97%
Chemical Name or Material (1-Thieno[3, 2-d]pyrimidin-4-ylpiperid-4-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'