missing translation for 'onlineSavingsMsg'
Learn More

1-(Trifluoroacetyl)imidazole, 98+%, Thermo Scientific Chemicals

Product Code. 11302675
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1546-79-8
Molecular Formula C5H3F3N2O
Molecular Weight (g/mol) 164.087
MDL Number MFCD00014501
InChI Key SINBGNJPYWNUQI-UHFFFAOYSA-N
Synonym 1-trifluoroacetyl imidazole, n-trifluoroacetylimidazole, 1-trifluoroacetyl-1h-imidazole, 1h-imidazole, 1-trifluoroacetyl, unii-8c6041ow3i, tfai, 2,2,2-trifluoro-1-1h-imidazol-1-yl ethanone, n-trifluoroacetyl imidazole, 2,2,2-trifluoro-1-imidazol-1-yl ethanone, 2,2,2-trifluoro-1-1h-imidazol-1-yl ethan-1-one
PubChem CID 73767
IUPAC Name 2,2,2-trifluoro-1-imidazol-1-ylethanone
SMILES C1=CN(C=N1)C(=O)C(F)(F)F

Specifications

Density 1.441
Boiling Point 45°C to 46°C (14 mmHg)
Flash Point 45°C (113°F)
Odor Pungent
Refractive Index 1.424
Quantity 10 g
UN Number UN1993
Beilstein 608904
Sensitivity Moisture sensitive; May form a precipitate
Formula Weight 164.09
Percent Purity ≥98%
Chemical Name or Material 1-(Trifluoroacetyl)imidazole
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(Trifluoroacetyl)imidazole, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.