missing translation for 'onlineSavingsMsg'
Learn More

1-(Trimethylsiloxy)cyclohexene, 98%, Thermo Scientific Chemicals

Product Code. 11433127
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6651-36-1
Molecular Formula C9H18OSi
Molecular Weight (g/mol) 170.33
MDL Number MFCD00001541
InChI Key SBEMOANGDSSPJY-UHFFFAOYSA-N
Synonym 1-trimethylsilyloxy cyclohexene, cyclohex-1-en-1-yloxy trimethylsilane, 1-trimethylsiloxy cyclohexene, silane, 1-cyclohexen-1-yloxy trimethyl, 1-cyclohexenyloxytrimethylsilane, 1-cyclohexen-1-yloxy trimethylsilane, cyclohexene, 1-trimethylsilyl oxy, cyclohexenyloxy trimethylsilane, 1-trimethylsiloxycyclohexene, cyclohexenyloxytrimethylsilane
PubChem CID 81161
IUPAC Name cyclohexen-1-yloxy(trimethyl)silane
SMILES C[Si](C)(C)OC1=CCCCC1

Specifications

Boiling Point 165°C
Quantity 10 g
UN Number UN1993
Beilstein 1859394
Flash Point 42°C (107°F)
Solubility Information Not miscible or difficult to mix.
Formula Weight 170.33
Percent Purity 98%
Refractive Index 1.447
Sensitivity Moisture sensitive
Density 0.883
Chemical Name or Material 1-(Trimethylsiloxy)cyclohexene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.