missing translation for 'onlineSavingsMsg'
Learn More

1-Trimethylsilyl-1-pentyne, 98%, Thermo Scientific Chemicals

Product Code. 15450218
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18270-17-2
Molecular Formula C8H16Si
Molecular Weight (g/mol) 140.30
MDL Number MFCD00077890
InChI Key CABCDUQQPBAHEE-UHFFFAOYSA-N
Synonym 1-trimethylsilyl-1-pentyne, trimethyl pent-1-yn-1-yl silane, trimethyl pent-1-ynyl silane, 1-pentyn-1-yltrimethylsilane, 1-tms-1-pentyne, acmc-209ejs, 1-trimethylsilylpent-1-yne, 1-trimethylsi;lyl-1-pentyne
PubChem CID 2760860
SMILES CCCC#C[Si](C)(C)C

Specifications

Boiling Point 135°C
Quantity 5 g
Linear Formula CH3CH2CH2C≡CSi(CH3)3
UN Number UN1993
Beilstein 1744218
Flash Point 23°C (73°F)
IUPAC Name trimethyl(pent-1-yn-1-yl)silane
Formula Weight 140.3
Percent Purity 98%
Refractive Index 1.425
Density 0.765
Chemical Name or Material 1-Trimethylsilyl-1-pentyne
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.