Learn More
Masterflex™ Stopcocks with Luer Connection, male lock, 1-way
Use these stopcocks for a variety of applications. Masterflex™ Stopcocks with Luer Connections feature a PC body and an HDPE stem.
Brand:  Masterflex™ 30600-05
Product Code. 11719709
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • One-way stopcock with male lock luer connection
  • Handle rotates 90 degrees on one-way stopcocks
Specifications
Specifications
Polycarbonate
100 Pack
100 Pack