missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ 10 to 8000 ml Dispensing Tool

Interchangeable stator and rotor that is flexible, easy to use, rapid, and user-friendly. Velp Scientifica™ 10 to 8000 ml Dispensing Tool is an instrument for a wide range of uses that ensures excellent performance and safety.

Brand:  Velp Scientifica™ A00000033

 View more versions of this product

Product Code. 15369524

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Can be assembled in a few seconds and without the use of tools
  • Consisting of a high-speed rotor with sharp blades lodged within a stationary stator with openings
  • Makes the OV5 suitable for the most diverse applications
Specifications

Specifications

Dispensing Tool
Rotor: 23 mm, Stator: 30 mm
220 mm
OV5 Homogenizer
1 Ea.
10 mL to 8 L
40 to 175 mm
Paper and Tissue Industry, Food Industry, Tobacco Industry, Medicine, Sewage Pollution Control applications
VSS3CSR3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.