missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wheaton™ 10 μL Inoculation Loop
Inoculation Loop for delivering a known volume of fluid sample to enable accurate microbiological analysis
73.05 € - 394.00 €
Specifications
Material | ABS |
---|---|
Sterility | Sterile |
Disposable | Single-use |
Color | Blue |
Packaging Type | Packaged in peel pouch |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
16415009
|
Wheaton™
711300 |
Short |
394.00 €
Pack of 6000 |
Estimated Shipment: 26-03-2025
Log in to see stock. |
|||||
16455009
|
Wheaton™
711420 |
Standard |
73.05 €
Pack of 500 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
Description
- For dispersing samples on agar surface
- Meet quantitative standards for management of microbiological samples
Specifications
ABS | |
Single-use | |
Packaged in peel pouch |
Sterile | |
Blue | |
Not Autoclavable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title