missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ 10 mL Urine Tube with Printed Label

Sterile container offering sample security and user safety with excellent leak free performance.

Brand:  Wheaton™ 327230

 View more versions of this product

Product Code. 16404969

  • 228.00 € / Pack of 450

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Wide container range to suit most user needs
  • With collection cup
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Red
Single-use
Sterile
EU Directive 98/79/EC
Urine analysis
Label, Collection Cup, Boric Acid
Yes
50 Pk.
Screw Cap
Polystyrene
Round
16 mm
100 mm
Printed
450/Cs.
10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.