Learn More
10 NTU Ratio Turbidity Standard, Reagecon
BIOPHARMA
A suspension of polymer microspheres designed to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards
Brand:  Reagecon CRSR-10-100
Product Code. 11576573
  • 118.00 € / 100mL
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Guaranteed stability throughout shelf life even after opening bottle
  • ± 1% accuracy
  • NIST traceable
  • US EPA approved
Specifications
Specifications
Turbidity Standard
Liquid
100 mL
Ratio Instruments
± 1% NTU
HDPE Bottle