missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Phenylanthracene-9-boronic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is used as pharmaceutical intermediate and as OLED materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate and as OLED materials.
Solubility
Soluble in methanol.
Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
It is used as pharmaceutical intermediate and as OLED materials.
Solubility
Soluble in methanol.
Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.

Specifications
Specifications
Quantity | 1 g |
Beilstein | 11434760 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in methanol. |
Formula Weight | 298.15 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 10-Phenylanthracene-9-boronic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction