missing translation for 'onlineSavingsMsg'
Learn More

10-Phenylanthracene-9-boronic acid, 98%

Product Code. 15442329
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15442329

Brand: Thermo Scientific Alfa Aesar H64911.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 334658-75-2 | C20H15BO2 | 298.15 g/mol

It is used as pharmaceutical intermediate and as OLED materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate and as OLED materials.

Solubility
Soluble in methanol.

Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334658-75-2
Molecular Formula C20H15BO2
Molecular Weight (g/mol) 298.15
MDL Number MFCD11111989
InChI Key RVPCPPWNSMAZKR-UHFFFAOYSA-N
Synonym 10-phenylanthracen-9-yl boronic acid, 10-phenyl-9-anthracene boronic acid, 10-phenyl-9-anthraceneboronic acid, 10-phenylanthracene-9-boronic acid, 10-phenylantrhacen-9-yl boronic acid, boronic acid, 10-phenyl-9-anthracenyl, pubchem19639, 9-borono-10-phenylanthracene, 10-phenylanthracene-9-ylboronic acid, 10-phenyl-9-anthryl boronic acid
PubChem CID 22247164
IUPAC Name (10-phenylanthracen-9-yl)boronic acid
SMILES OB(O)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12

Specifications

Quantity 250 mg
Beilstein 11434760
Sensitivity Light sensitive
Solubility Information Soluble in methanol.
Formula Weight 298.15
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 10-Phenylanthracene-9-boronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.