Learn More
Megro™ 100/125mL Urine Beaker with Green Screw Cap
Ideal for the collection and storage of samples of all types.
Brand:  Megro™ MEG122500
Product Code. 17800260
  • 113.54 € / Pack of 500
Explore more special offers
Description
Description
  • Pressure resistant and shatterproof, solid and steady
  • Optimal visibility of contents, graduated
  • Wide opening for easy and safe filling
  • Fluid-tight sealing screw cap with thread
  • Sealing is completely uncomplicated and immediately comprehensible
  • Frosted surface on cups and lids for identification marking
  • Made of polypropylene/HDPE
  • 20 x 25 beakers and 5 x 100 caps