missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMS™ Series 3 Model 100WA Cooled Incubator, 100 L, Stainless Steel
Temperature Range: -10°C to +50°C
Brand: LMS™ 100A
This item is not returnable.
View return policy
Description
- Range of mid-sized benchtop and freestanding cooled incubators with a white stove enamel mild steel exterior, foam insulation and stainless steel interiors
- Reliable and accurate temperature control
- Standard single and dual temperature, plus programmable models available
- Temperature range -10.0°C to +50.0°C, temperature variation with average workload±0.5°C
- Self-evaporating condensate drip tray
- White plastic coated wire shelves
- Energy efficient control system and eco-friendly
- P.I.D. heating and cooling to ensure stable internal temperature
- Pre-set safety cutout
- Hermetically sealed refrigeration system
- Door lock
- Digital controls are easy to read and simple to use
- Full range offactory-fitted optional extras available, including LED lighting, access porthole, alarm system, inner perspex door, internal socket and chart recorder
- Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests
Specifications
ISO 9001:2008 | |
Freestanding incubator | |
Stainless Steel | |
500 mm x 380 mm | |
Stove Enameled Mild Steel | |
-10°C to +50°C | |
Digital | |
100WA | |
CFC-free | |
±0.1°C | |
2 Years | |
21.3 in. | |
32.3 in. | |
820 mm | |
17.3 in. | |
50 kg | |
230 V |
Dual temperature | |
100 L | |
Fan assisted air circulation via premixing chamber | |
Optional Fluorescent/UV | |
2 | |
Refrigerated Incubator | |
Foamed polyurethane | |
1 | |
435 x 270 mm | |
±0.5°C | |
24.0 in. | |
540 mm | |
19.3 in. | |
490 mm | |
440 mm | |
240 V, 50 Hz | |
50 Hz |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction